Combination Blocks
STT-W14-COMBO block, Ø 8,3/13,9/26,2 mm, 6/12/13/25 mm round-bottom tubes, 6/5/3, 48,4 mm
Katalognr. VELPA00000511
New Product
Anbieter: VELP SCIENTIFIC
Direkt zu
Product Details & Documents
Technische Daten
- No. of Holes:6/5/3
- Description:STT-W14-COMBO block
- Seq:1
- Depth:48,4 mm
- Well size:Ø 8,3/13,9/26,2 mm
- For:6/12/13/25 mm round-bottom tubes