Orders LinkContactUsLinkComponent
Combination Blocks | Avantor
Combination Blocks
MCT-W21-COMBO block, Ø 7,9/11,1/11,5 mm, 0,5/1,5/2,0 ml microcentrifuge tubes, 6/10/5, 27,6/39,1/38,1 mm
Katalognr. VELPA00000513
New Product
Anbieter:   VELP SCIENTIFIC
Combination Blocks
Katalognr. VELPA00000513
Anbieter:  VELP SCIENTIFIC
New Product
Combination Blocks
Combination Blocks
MCT-W21-COMBO block, Ø 7,9/11,1/11,5 mm, 0,5/1,5/2,0 ml microcentrifuge tubes, 6/10/5, 27,6/39,1/38,1 mm
Preis
SFr. 170.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • No. of Holes:
    6/10/5
  • Description:
    MCT-W21-COMBO block
  • Seq:
    3
  • Depth:
    27,6/39,1/38,1 mm
  • Well size:
    Ø 7,9/11,1/11,5 mm
  • For:
    0,5/1,5/2,0 ml microcentrifuge tubes