Methyl-β-D-glucopyranoside
Bottle
Catalog # APOSBIM8001-100G
New Product
Supplier: Apollo Scientific
Methyl-β-D-glucopyranoside
Catalog # APOSBIM8001-100G
Supplier: Apollo Scientific
New Product
CAS Number: 709-50-2
Jump to:
Product Details & Documents
Synonyms: Methyl β-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
- CAS Number:709-50-2
- Formula:C₇H₁₄O₆
- MDL Number:MFCD00006602
- Molecular Weight:194.1825 g/mol
- Melting Pt:111…113 °C
- Storage Temperature:Freezer
- EINECS:211-909-9
Specifications
Specifications
- Pack type:Bottle



