Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Copper

Copper

Bottle
Catalog # APOSIN1526-500G
New Product
Supplier:   Apollo Scientific
Copper
Catalog # APOSIN1526-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  
Copper

Bottle

CAS Number:  
Price
SFr. 2’140.00
Log in for availability
1 * 500 g
Product Details & Documents
  • Molecular Weight:63,546 g/mol
  • UN:3089
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.