Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Copper 99,9%, nanopowder 30 - 50 nm

Copper 99,9%, nanopowder 30 - 50 nm

Bottle
Catalog # APOSIN1526-5G
New Product
Supplier:   Apollo Scientific
Copper 99,9%, nanopowder 30 - 50 nm
Catalog # APOSIN1526-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  7440-50-8
Copper 99,9%, nanopowder 30 - 50 nm

Bottle

CAS Number:  7440-50-8
Price
SFr. 55.50
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Inorganic salt or catalyst

  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63,546 g/mol
  • UN:3089
Specifications

Specifications

  • Pack type:
    Bottle

Specifications