Chlorophenol red
Bottle
Catalog # APOSBI4558-1G
New Product
Supplier: Apollo Scientific
Chlorophenol red
Catalog # APOSBI4558-1G
Supplier: Apollo Scientific
New Product
CAS Number: 4430-20-0
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Number:4430-20-0
- Formula:C₁₉H₁₂Cl₂O₅S
- MDL Number:MFCD00005877
- Molecular Weight:423,273 g/mol
- EINECS:224-619-2
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



