m-PEG4-amine
Bottle
Catalog # APOSBIPG1582-5G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3,6,9,12-Tetraoxatridecanamine
- CAS Number:85030-56-4
- Formula:C₉H₂₁NO₄
- Molecular Weight:207.27 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



