Cellulose Maru Xin type, microcrystalline
Bottle
Catalog # APOSOR59788-25G
New Product
Supplier: Apollo Scientific
Cellulose Maru Xin type, microcrystalline
Catalog # APOSOR59788-25G
Supplier: Apollo Scientific
New Product
CAS Number: 9004-34-6
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Microcrystallinecellulose
- CAS Number:9004-34-6
- MDL Number:MFCD00081512
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



