Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cholesterol 95%

Cholesterol 95%

Bottle
Catalog # APOSOR1030208-2.5K
Irritant
New Product
Supplier:   Apollo Scientific
Cholesterol 95%
Catalog # APOSOR1030208-2.5K
Supplier:  Apollo Scientific
New Product
CAS Number:  57-88-5
Cholesterol 95%

Bottle

CAS Number:  57-88-5
Price
SFr. 998.00
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
  • CAS Number:57-88-5
  • Formula:C₂₇H₄₆O
  • MDL Number:MFCD00003646
  • Molecular Weight:386,55 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications