α-Cyclodextrin
Bottle
Catalog # APOSBIB6038-500G
New Product
Supplier: Apollo Scientific
α-Cyclodextrin
Catalog # APOSBIB6038-500G
Supplier: Apollo Scientific
New Product
CAS Number: 10016-20-3
Jump to:
Product Details & Documents
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.84 g/mol
- Melting Pt:>278 °C
- Melting Pt:>278 °C
- Storage Temperature:Ambient
- EINECS:233-007-4
Specifications
Specifications
- Pack type:Bottle



