Methotrexate
Bottle
APOSBIM0130-25G
New Product
An antineoplastic agent which acts as an antimetaboliteof folic acid.
: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- :59-05-2
- :C₂₀H₂₂N₈O₅
- :MFCD00150847
- :454.44 g/mol
- :185…204 °C
- :Freezer
- :1544
- :6.1,III
- Pack type:Bottle