Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Actarit 98%

Actarit 98%

Bottle
Catalog # APOSOR911806-1G
New Product
Supplier:   Apollo Scientific
Actarit 98%
Catalog # APOSOR911806-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  18699-02-0
Actarit 98%

Bottle

CAS Number:  18699-02-0
Price
SFr. 29.50
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
  • CAS Number:18699-02-0
  • Formula:C₁₀H₁₁NO₃
  • MDL Number:MFCD00196165
  • Molecular Weight:193.20 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.