Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2',3'-Dimethoxyacetophenone 95%

2',3'-Dimethoxyacetophenone 95%

Bottle
Catalog # APOSOR938335-100G
New Product
Supplier:   Apollo Scientific
2',3'-Dimethoxyacetophenone 95%
Catalog # APOSOR938335-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  38480-94-3
2',3'-Dimethoxyacetophenone 95%

Bottle

CAS Number:  38480-94-3
Price
SFr. 2’560.00
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:38480-94-3
  • Formula:C₁₀H₁₂O₃
  • MDL Number:MFCD03424412
  • Molecular Weight:180.2005 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications