Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2',3'-Dimethoxyacetophenone 95%

2',3'-Dimethoxyacetophenone 95%

Bottle
Catalog # APOSOR938335-5G
New Product
Supplier:   Apollo Scientific
2',3'-Dimethoxyacetophenone 95%
Catalog # APOSOR938335-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  38480-94-3
2',3'-Dimethoxyacetophenone 95%

Bottle

CAS Number:  38480-94-3
Price
SFr. 139.00
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:38480-94-3
  • Formula:C₁₀H₁₂O₃
  • MDL Number:MFCD03424412
  • Molecular Weight:180.2005 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.