2',3'-Dimethoxyacetophenone 95%
Bottle
Katalognr. APOSOR938335-250MG
New Product
Anbieter: Apollo Scientific
2',3'-Dimethoxyacetophenone 95%
Katalognr. APOSOR938335-250MG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 38480-94-3
Direkt zu
Product Details & Documents
- CAS Nummer:38480-94-3
- Formel:C₁₀H₁₂O₃
- MDL-Nummer:MFCD03424412
- Molekulargewicht:180.2005 g/mol
- Lagertemperatur:Ambient
Technische Daten
Technische Daten
- Pack type:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



