Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Uridine ≥98%

Bottle
Catalog # APOSBUP16801-10G
New Product
Supplier:   Apollo Scientific
Uridine ≥98%
Catalog # APOSBUP16801-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  58-96-8
Uridine ≥98%

Bottle

CAS Number:  58-96-8
Price
SFr. 633.00
Log in for availability
1 * 10 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • Molecular Weight:244,2 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications