Uridine ≥98%
Bottle
Catalog # APOSBUP16801-500MG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
- CAS Number:58-96-8
- Formula:C₉H₁₂N₂O₆
- Molecular Weight:244,2 g/mol
Specifications
Specifications
- Pack type:Bottle



