Order Entry
Switzerland
Orders LinkContactUsLinkComponent
AMP-945 ≥98%

AMP-945 ≥98%

Bottle
Catalog # APOSBUP07951-50MG
New Product
Supplier:   Apollo Scientific
AMP-945 ≥98%
Catalog # APOSBUP07951-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1393653-34-3
AMP-945 ≥98%

Bottle

CAS Number:  1393653-34-3
Price
SFr. 1’220.00
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Narmafotinib
  • CAS Number:1393653-34-3
  • Formula:C₂₈H₃₂F₃N₅O₂
  • Molecular Weight:527,58 g/mol
Specifications

Specifications

  • Pack type:
    Bottle
  • Cat. No.:
    APOSBUP07951-50MG

Specifications