Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Actarit ≥98%

Actarit ≥98%

Bottle
Catalog # APOSBUP17989-1G
New Product
Supplier:   Apollo Scientific
Actarit ≥98%
Catalog # APOSBUP17989-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  18699-02-0
Actarit ≥98%

Bottle

CAS Number:  18699-02-0
Price
SFr. 108.00
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
  • CAS Number:18699-02-0
  • Formula:C₁₀H₁₁NO₃
  • Molecular Weight:193,2 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications