Actarit ≥98%
Bottle
Catalog # APOSBUP17989-200MG
New Product
Supplier: Apollo Scientific
Actarit ≥98%
Catalog # APOSBUP17989-200MG
Supplier: Apollo Scientific
New Product
CAS Number: 18699-02-0
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: 2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
- CAS Number:18699-02-0
- Formula:C₁₀H₁₁NO₃
- Molecular Weight:193,2 g/mol
Specifications
Specifications
- Pack type:Bottle



