Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Decyl aldehyde ≥95%

Decyl aldehyde ≥95%

Bottle
Catalog # APOSBUP18889-10G
New Product
Supplier:   Apollo Scientific
Decyl aldehyde ≥95%
Catalog # APOSBUP18889-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  112-31-2
Decyl aldehyde ≥95%

Bottle

CAS Number:  112-31-2
Price
SFr. 122.00
Log in for availability
1 * 10 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Decylaldehyde, Decanal, Decyl aldehyde
  • CAS Number:112-31-2
  • Formula:C₁₀H₂₀O
  • Molecular Weight:156,27 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.