Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Decyl aldehyde ≥95%

Decyl aldehyde ≥95%

Bottle
Catalog # APOSBUP18889-1G
New Product
Supplier:   Apollo Scientific
Decyl aldehyde ≥95%
Catalog # APOSBUP18889-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  112-31-2
Decyl aldehyde ≥95%

Bottle

CAS Number:  112-31-2
Price
SFr. 46.80
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Decylaldehyde, Decanal, Decyl aldehyde
  • CAS Number:112-31-2
  • Formula:C₁₀H₂₀O
  • Molecular Weight:156,27 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications