Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methotrexate ≥95%

Methotrexate ≥95%

Bottle
Catalog # APOSBUP17033-500MG
New Product
Supplier:   Apollo Scientific
Methotrexate ≥95%
Catalog # APOSBUP17033-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  59-05-2
Methotrexate ≥95%

Bottle

CAS Number:  59-05-2
Price
SFr. 217.00
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
  • CAS Number:59-05-2
  • Formula:C₂₀H₂₂N₈O₅
  • Molecular Weight:454,44 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications