Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Lanolin 98%

Lanolin 98%

Bottle
Catalog # APOSOR1024474-100G
Irritant
New Product
Supplier:   Apollo Scientific
Lanolin 98%
Catalog # APOSOR1024474-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  8006-54-0
Lanolin 98%

Bottle

CAS Number:  8006-54-0
Price
SFr. 26.00
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • EINECS:232-348-6
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.