Orders LinkContactUsLinkComponent
Intelsius® Pathopouch
New Product
Supplier:   DGP INTELSIUS
Intelsius® Pathopouch
Supplier:  DGP INTELSIUS
New Product
Certificates
Intelsius® Pathopouch
Intelsius® Pathopouch
2 Options Available Below
Product Details & Documents
The Intelsius® Pathopouch range are 95 kPa flexible pressure vessels designed to meet with transport regulations.
  • 95 kPa plastic secondary packaging
  • Outer pouch for paperwork
  • Leak-proof
They are leak-proof and suitable to hold a wide range of primary containers, such as blood vials, specimen containers, swabs, blood collection bags and medical devices.
Certifications: ISTA Certified Class 6.2 Category A & B
Product Family Options
Product Information
  • Size
    L×W
    Availability
    Price
  • DGINBP003
    95 kPa pouch for pathology
    95 kPa pouch for pathology
    A4
    308×230 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    New Product
    New Product
  • DGINBP005
    95 kPa pouch for pathology
    95 kPa pouch for pathology
    A3
    359×261 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    New Product
    New Product