Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Lanolin

Lanolin

Bottle
Catalog # APOSBUP18167-500MG
New Product
Supplier:   Apollo Scientific
Lanolin
Catalog # APOSBUP18167-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  8006-54-0
Lanolin

Bottle

CAS Number:  8006-54-0
Price
SFr. 60.70
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • Formula:C₃₄H₆₈O₂
  • Molecular Weight:508,92 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.