Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-Ribose 99%

D-Ribose 99%

New Product
Supplier:   Apollo Scientific
D-Ribose 99%
Supplier:  Apollo Scientific
New Product
Certificates

4 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

  • Store at 2 to 8 °C
  • Non-hazardous
  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150,1299 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-059-4
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 26.00
    SFr. 26.00
    1 * 25 g
       SFr. 26.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 29.50
    SFr. 29.50
    1 * 100 g
       SFr. 29.50

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 69.30
    SFr. 69.30
    1 * 500 g
       SFr. 69.30

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - SFr. 304.00
    SFr. 304.00
    1 * 2,5 kg
       SFr. 304.00

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.