Chlorophenol red
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Number:4430-20-0
- Formula:C₁₉H₁₂Cl₂O₅S
- MDL Number:MFCD00005877
- Molecular Weight:423,273 g/mol
- EINECS:224-619-2
3 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Number:4430-20-0
- Formula:C₁₉H₁₂Cl₂O₅S
- MDL Number:MFCD00005877
- Molecular Weight:423,273 g/mol
- EINECS:224-619-2



