Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Lanolin 98%

Lanolin 98%

Irritant
New Product
Supplier:   Apollo Scientific
Lanolin 98%
Supplier:  Apollo Scientific
New Product
Certificates

2 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • EINECS:232-348-6
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 26.00
    SFr. 26.00
    1 * 100 g
       SFr. 26.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 69.30
    SFr. 69.30
    1 * 500 g
       SFr. 69.30

    Specifications:
    New Product
    New Product