Bromothymol Blue
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent
- Non-hazardous
Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-59-5
- Formula:C₂₇H₂₈Br₂O₅S
- MDL Number:MFCD00005872
- Molecular Weight:624,38 g/mol
- EINECS:200-971-2
5 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent
- Non-hazardous
Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-59-5
- Formula:C₂₇H₂₈Br₂O₅S
- MDL Number:MFCD00005872
- Molecular Weight:624,38 g/mol
- EINECS:200-971-2
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



