Actarit ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/life science applications
Synonyms: 2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
- CAS Number:18699-02-0
- Formula:C₁₀H₁₁NO₃
- Molecular Weight:193,2 g/mol
3 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: 2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
- CAS Number:18699-02-0
- Formula:C₁₀H₁₁NO₃
- Molecular Weight:193,2 g/mol
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



