Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(-)-Limonene ≥95%

(-)-Limonene ≥95%

New Product
Supplier:   Apollo Scientific
(-)-Limonene ≥95%
Supplier:  Apollo Scientific
New Product
Certificates
(-)-Limonene ≥95%

3 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene
  • CAS Number:5989-54-8
  • Formula:C₁₀H₁₆
  • Molecular Weight:136,23 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 53.70
    SFr. 53.70
    1 * 200 mg
       SFr. 53.70

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - SFr. 84.90
    SFr. 84.90
    1 * 500 mg
       SFr. 84.90

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 123.00
    SFr. 123.00
    1 * 1 g
       SFr. 123.00

    Specifications:
    New Product
    New Product