NMR Spinners
NMR spinner designed for 3 and 5 mm NMR tubes, compatible with standardbore NMR systems. Available in different materials (POM, Kel-F) to suit a wide range of experimental temperatures.
POM: 0 to 80 °C
Kel-F: −40 to +120 °C
These spinners ensure stable rotation, precise tube positioning, and reliable performance across a wide range of NMR applications.
NMR spinner designed for 3 and 5 mm NMR tubes, compatible with standardbore NMR systems. Available in different materials (POM, Kel-F) to suit a wide range of experimental temperatures.
POM: 0 to 80 °C
Kel-F: −40 to +120 °C
These spinners ensure stable rotation, precise tube positioning, and reliable performance across a wide range of NMR applications.
Product Family Options
- DescriptionØ
- New ProductNew Product
- New ProductNew Product