Orders LinkContactUsLinkComponent
EasyPlus™ Titration Electrodes
EasyPlus™ Titration Electrodes
New Product
Supplier:   Mettler - Toledo
EasyPlus™ Titration Electrodes
Supplier:  Mettler - Toledo
New Product
Certificates
EasyPlus™ Titration Electrodes
EasyPlus™ Titration Electrodes
5 Options Available Below
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus titrators only.
Electrode only.
Product Family Options
Product Information
  • Range
    For
    Availability
    Price
  • METT30043103
    EG11-BNC sensor pH aqueous
    EG11-BNC sensor pH aqueous
    ±2000 mV/0 - 14 pH
    Easy Pro, Easy EPM, Easy pH titrators
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 191.50
    SFr. 191.50
    1 * 1 ST
       SFr. 191.50
    Specifications:
    New Product
    New Product
  • METT30043106
    EG13-BNC sensor pH non-aqueous
    EG13-BNC sensor pH non-aqueous
    ±2000 mV/0 - 14 pH
    Easy Pro, Easy EPM, Easy pH titrators
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 206.00
    SFr. 206.00
    1 * 1 ST
       SFr. 206.00
    Specifications:
    New Product
    New Product
  • METT30043107
    EM45-BNC sensor precipitation
    EM45-BNC sensor precipitation
    ±2000 mV/0 - 14 pH
    Easy Pro, Easy Cl titrators
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 198.50
    SFr. 198.50
    1 * 1 ST
       SFr. 198.50
    Specifications:
    New Product
    New Product
  • METT30043105
    EM40-BNC sensor redox mV
    EM40-BNC sensor redox mV
    ±2000 mV
    Easy Pro, Easy Ox titrators
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 236.00
    SFr. 236.00
    1 * 1 ST
       SFr. 236.00
    Specifications:
    New Product
    New Product
  • METT30043104
    EM43-BNC sensor redox Ipol
    EM43-BNC sensor redox Ipol
    0 - 2000 mV
    Easy Pro, Easy KFV, Easy Ox titrators
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 264.00
    SFr. 264.00
    1 * 1 ST
       SFr. 264.00
    Specifications:
    New Product