Blocks for COD
New Product
Supplier: VELP SCIENTIFIC
Jump to:
About this item
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
3 Options Available Below
Product Details & Documents
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
Product Family Options
Product Information
- Well sizeForNo. of HolesDepthAvailabilityPrice
- 705-1207COD-W04-D420-A blockCOD-W04-D420-A blockØ 42,0 mm42 mm round-bottom tubes451 mmLog in for availability1 * 1 STSFr. 170.00Specifications:New ProductNew Product
- 705-1208COD-W20-COMBO blockCOD-W20-COMBO blockØ 16,0/22,0 mm16/22 mm round-bottom tubes18/247/45,5 mmLog in for availability1 * 1 STSFr. 170.00Specifications:New ProductNew Product
- 705-1206COD-W24-D160-A blockCOD-W24-D160-A blockØ 16,0 mm16 mm round-bottom tubes2447 mmLog in for availability1 * 1 STSFr. 170.00Specifications:New ProductNew Product