Orders LinkContactUsLinkComponent
Combination Blocks
Combination Blocks
New Product
Supplier:   VELP SCIENTIFIC
Combination Blocks
Supplier:  VELP SCIENTIFIC
New Product
Certificates
Combination Blocks
Combination Blocks
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Well size
    For
    No. of Holes
    Depth
    Availability
    Price
  • VELPA00000511
    STT-W14-COMBO block
    STT-W14-COMBO block
    Ø 8,3/13,9/26,2 mm
    6/12/13/25 mm round-bottom tubes
    6/5/3
    48,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 170.00
    SFr. 170.00
    1 * 1 ST
       SFr. 170.00
    Specifications:
    New Product
    New Product
  • VELPA00000512
    CCT-W09-COMBO block
    CCT-W09-COMBO block
    Ø 11,1/17,1/29 mm
    1,5/15/50 ml conical centrifuge
    4/3/2
    39,1/44,5/47,6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 170.00
    SFr. 170.00
    1 * 1 ST
       SFr. 170.00
    Specifications:
    New Product
    New Product
  • VELPA00000513
    MCT-W21-COMBO block
    MCT-W21-COMBO block
    Ø 7,9/11,1/11,5 mm
    0,5/1,5/2,0 ml microcentrifuge tubes
    6/10/5
    27,6/39,1/38,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 170.00
    SFr. 170.00
    1 * 1 ST
       SFr. 170.00
    Specifications:
    New Product