Orders LinkContactUsLinkComponent
External Probe Adapter
External adapter that provides access to an external probe CAP-MP (Ø 3 - 5 mm), for continuous validation processes
  ESPICAP-MP
 :   RAYPA
External Probe Adapter
  ESPICAP-MP
 :  RAYPA
External Probe Adapter
External Probe Adapter
External adapter that provides access to an external probe CAP-MP (Ø 3 - 5 mm), for continuous validation processes
1 * 1 ST
Allows access for an external probe to obtain a temperature reading from the culture medium, independent of the reading taken by the equipment's flexible probe.
  • The port is located on the door of the media preparator
  • The TP-VAL-MP temperature probe, compatible for this port, is available upon request
  • The external probe adapter is composed of three parts: stainless steel gland, cylindrical chrome-plated brass extension and polypropylene GL-32 cap
  • Description:
    External adapter that provides access to an external probe CAP-MP (Ø 3 - 5 mm), for continuous validation processes