Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Hydrocortisone ≥99% Adrenal glucocorticoid

Hydrocortisone ≥99% Adrenal glucocorticoid

Marketsource
Supplier:   Abcam
Hydrocortisone ≥99% Adrenal glucocorticoid
Supplier:  Abcam
Marketsource
Certificates

2 Options Available Below

Product Details & Documents

Adrenal glucocorticoid. Stimulates gluconeogenesis, activates anxiolytic and anti-infla mMatory pathways.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:60103-17-5
  • Molecular Weight:362,5 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Hydrocortisone
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 64.90
    SFr. 64.90
    1 * 1 g
       SFr. 64.90

    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 170.00
    SFr. 170.00
    1 * 5 g
       SFr. 170.00

    Specifications:
    Marketsource
    Marketsource