Tridecanoic acid ≥99% Phospholipase A2 substrate
Catalog # ABCAAB143889-100
Marketsource
Supplier: Abcam
Tridecanoic acid ≥99% Phospholipase A2 substrate
Catalog # ABCAAB143889-100
Supplier: Abcam
Marketsource
CAS Number: 638-53-9
Jump to:
Product Details & Documents
Phospholipase A2 substrate. 13-carbon saturated fatty acid. Product of anaerobic biodegradation of n-hexadecane.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Synonyms: n-Tridecanoic acid, Tridecylic acid
- CAS Number:638-53-9
- Molecular Weight:214,34 g/mol
Specifications
Specification Test Results
- Constituents::100% Tridecanoic acid



