Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Native lysis Buffer, Abcam

Native lysis Buffer, Abcam

Native lysis Buffer
Catalog # ABCAAB156035-15
Supplier:   Abcam
Native lysis Buffer, Abcam
Catalog # ABCAAB156035-15
Supplier:  Abcam
CAS Number:  
Native lysis Buffer, Abcam

Native lysis Buffer

CAS Number:  
Price
SFr. 54.90
Log in for availability
1 * 15 mL
Product Details & Documents

Abcam’s 1X Native lysis buffer is an efficient means of cell lysis and protein solubilization for both adherent and suspension cultured mammalian cells.

  • pH: 7.5 Constituents: 1.5% Dodecyl maltoside, 0.6% HEPES, 0.58% Sodium chloride, 0.11% Sodium pyrophosphate decahydrate, 0.04% EGTA, 0.03% EDTA, 0.02% Sodium orthovanadate, 0.02% disodium;1,3-dihydroxypropan-2-yl phosphate;hydrate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Native lysis Buffer
  • Cat. No.:
    ABCAAB156035-15
  • Storage temperature:
    Store at +4°C.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.