Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cell Lysis Buffer, Abcam

Cell Lysis Buffer, Abcam

Cell Lysis Buffer
Catalog # ABCAAB152163-15
Marketsource
Supplier:   Abcam
Cell Lysis Buffer, Abcam
Catalog # ABCAAB152163-15
Supplier:  Abcam
Marketsource
CAS Number:  
Cell Lysis Buffer, Abcam

Cell Lysis Buffer

CAS Number:  
Price
SFr. 145.00
Log in for availability
1 * 15 mL
Product Details & Documents

ab152163 is a 10X Cell Lysis Buffer used to lyse cells under non-denaturing conditions.

  • pH: 7.5 Constituents: 10% Triton X-100, 8.8% Sodium chloride, 3.15% Tris HCl, 1.12% Sodium pyrophosphate decahydrate, 0.38% EGTA, 0.29% Tetrasodium ethylene diamine tetraacetate, 0.216% disodium;1,3-dihydroxypropan-2-yl phosphate;hydrate, 0.19% Sodium orthovanadate, 0.001% Leupeptin

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Cell Lysis Buffer
  • Cat. No.:
    ABCAAB152163-15
  • Storage temperature:
    Store at -20°C.

Specifications