Orders LinkContactUsLinkComponent
G-Biosciences TBE-Urea Sample Buffer
G-Biosciences TBE-Urea Sample Buffer
TBE-urea sample buffer, 30 mL
Catalog # 786-474
Supplier:   G-Biosciences
G-Biosciences TBE-Urea Sample Buffer
Catalog # 786-474
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences TBE-Urea Sample Buffer
G-Biosciences TBE-Urea Sample Buffer
TBE-urea sample buffer, 30 mL
CAS Number:  
Price
SFr. 35.30
Log in for availability
1 * 30 mL
Product Details & Documents
Loading buffer to resolve single-stranded DNA oligos or RNA into sharp, distinct bands.
  • Ready-to-use format: Pre-mixed buffer simplifies sample preparation and improves reproducibility
Broad application range: Used in molecular biology protocols involving RNA electrophoresis, ribonuclease protection assays, and sequencing gels.
Specifications
Specifications
  • Pk:
    30 mL
  • Description:
    TBE-urea sample buffer