G-Biosciences MOPS Running Buffers (10X)
Supplier: G-Biosciences
Certificates
About this item
A zwitterionic buffer providing consistent buffering capacity in the neutral range, ideal for RNA electrophoresis.
200 mM MOPS (pH 7,0), 80 mM sodium acetate, 10 mM EDTA
Convenient stock solution for dilution, saving prep time and storage space.
Frequently Bought Together