Proteometer-CV Consumables Kit (240)
Catalog # NOVIKIT-PCV-005-01
Supplier: NOVILYTIC
New Product
CAS Number:
Specifications
- Description:Proteometer-CV consumables kit
Specifications
About this item
Kit is for when customers needs more consumables but not the MD Unit or reactor.
- No sample preparation required - analysis directly from clarified fermentation broth without protein A purification
- Speed and efficiency - CV analysis in under 25 minutes
- Simplified workflow - all the components you need for seamless operation
- Exclusive technology - the only solution that performs charge variant analysis directly in clarified fermentation broth
- Store at −20 °C.Glassware not included
The Proteometer-CV kit offers an alternative to traditional charge variant methods that require protein A purification, thereby saving time and money. We are the only solution that performs charge variant analysis of samples directly in clarified fermentation broth, minimizing sample handling required for other charge variant analyses.
Reagent to be stored at −20 °C.
Accessories information: Reagent to be stored at −20 °C.
Ordering information:
Glassware not included. This kit does not include a Proteometer reactor.
Delivery information:
Includes 4× Proteometer-CV fluorescent reagent vias, 16× Prot-CV MD buffer packets, 20× Prot-CV MPA salt packets, 4× Prot-CV MPB salt packets, 1× reconstitution reagent - allows for analysis up to 240 samples.
Packaging: Supplied in a custom box with instructions.