ContactUsLinkComponent
Proteometer-UFT/CV Tubing Union
Proteometer-UFT/CV Tubing Union
Catalog # NOVIZDV-PFV-001
Supplier:  NOVILYTIC
New Product
Proteometer-UFT/CV Tubing Union
Catalog # NOVIZDV-PFV-001
Supplier:  NOVILYTIC
New Product
Specifications
  • Description:
    Proteometer-UFT/CV tubing union
Specifications
About this item
Replacement tubing union for use with the Proteometer UFT or CV kits. The UFT kit requires two unions. The CV Kit requires one.
  • Replacement tubing unions for use with the Proteometer-UFT and/or -CV kits
Intended for use with the Proteometer-UFT and Proteometer-CV kits. Reorder individually as needed.
Packaging: Supplied in a custom box with instructions.