Order Entry
Switzerland
ContactUsLinkComponent
 
Proteometer-L Fluorescent Reagent
Catalog # NOVIRGN-PL-0017
Supplier:  NOVILYTIC
New Product
CAS Number:  
undefined
Proteometer-L Fluorescent Reagent
Catalog # NOVIRGN-PL-0017
Supplier:  NOVILYTIC
New Product
CAS Number:  

Specifications

  • Description:
    Proteometer-L fluorescent reagent, 7 vials of the Proteometer-L fluorescent reagent

Specifications

About this item

Set of 7 reagent vials for use with the Proteometer-L kit.

  • Replacement vials for frequent users of the Proteometer-L kit
  • Store at −20 °C

Intended for use with the Proteometer-L kit. Crimped, sealed, sold in sets of seven.

Packaging: Supplied in a custom box with instructions.