ContactUsLinkComponent
Peha® Instrument Operating Scissor
Peha® Instrument Operating Scissor
Catalog # HART991082
Supplier:  HARTMANN PAUL
New Product
Peha® Instrument Operating Scissor
Catalog # HART991082
Supplier:  HARTMANN PAUL
New Product
Specifications
  • Type:
    Blunt/blunt straight
  • Length:
    145 mm
  • Pk:
    1
Specifications
About this item
Suitable for cutting threads and dressing materials, incisions as well as cutting or preparing tissue.
  • Single-use surgical steel instruments
    Devices are intended to be used in a sterile condition
Peha–instrument scissors are intended for transient treatment in the context of medical interventions used by healthcare professionals.
Packaging:
Pack of one.