Order Entry
Switzerland
ContactUsLinkComponent
VWR® PFAS Kits, 1,5 ml PP Vials with 9 mm Short Thread Cap, Silicone/Polypropylene
VWR® PFAS Kits, 1,5 ml PP Vials with 9 mm Short Thread Cap, Silicone/Polypropylene
Supplier:  VWR Collection
Certificates
undefined
VWR® PFAS Kits, 1,5 ml PP Vials with 9 mm Short Thread Cap, Silicone/Polypropylene
Supplier:  VWR Collection
Certificates

Specifications

  • Catalog No:
  • 548-7000A
  • 548-7005A
  • Kit:
  • KIT
  • KIT
  • Component vial:
  • 548-7002A (kits 548-7000A, 548-7005A)
  • 548-7002A (kits 548-7000A, 548-7005A)
  • Component cap:
  • 548-7003A (kit 548-7000A); 548-7012A (kit 548-7005A)
  • 548-7003A (kit 548-7000A); 548-7012A (kit 548-7005A)
  • Height with cap:
  • 5,7 mm
  • 5,7 mm
  • Ext.Ø with cap:
  • 11,5 mm
  • 11,5 mm

Specifications

Frequently Bought Together