ContactUsLinkComponent
EZ-Pak Filters
EZ-Pak Filters
  MILXEZHABG504
 :  Merck
New Product
EZ-Pak Filters
  MILXEZHABG504
 :  Merck
New Product
  • Ø:
    50 mm
  • Pore size:
    0,45 µm
  • Description:
    EZ-Pak filters black, gridded
  • Seq:
    2
EZ-Pak membranes and dispenser revolutionise filter handling for MF method testing by combining the benefits of ready-to-use, sterile, individually-packed membranes with unique handling ease. From convenient, ready-to-use test kits to individual tools, simply select the items that are right for you.
Applications: Clinical, cosmetics, environmental, food and beverage, industrial QC, pharmaceutical