Order Entry
Switzerland
ContactUsLinkComponent
SPINeasy® DNA Kit for Yeast
Catalog # 116557050.
Supplier:  MP Biomedicals
New Product
CAS Number:  
SPINeasy® DNA Kit for Yeast
Catalog # 116557050.
Supplier:  MP Biomedicals
New Product
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA kit for yeast
  • Storage temperature:
    Store at room temperature (15...30 °C), dry environment
  • No. of reactions:
    50 preps

Specifications

About this item

SPINeasy® DNA Kit for Yeast is a high-performance genomic DNA (gDNA) extraction kit which is developed based on silica-membrane spin-column technology. This kit enables quick isolation of gDNA from yeast cells, typically in less than 30 minutes.

  • For PCR amplification, restriction enzyme digestion, and sequencing

Yeast cell walls are resilient, hence it can become a challenge when trying to extract the intact purified DNA. Many times, the extracted DNA can be damaged or contaminated due to unintuitive workflow.

Provided in the kit, Column S and kit buffers are designed to deliver gDNA extracts of high yield and purity; the extracted gDNA is compatible with downstream applications such as PCR amplification, restriction enzyme digestion, and sequencing.

Components: Lysing matrix YG (50×2,0 ml), lysis buffer Y (30 ml), binding buffer Y (30 ml), wash buffer Y1 (32 ml), wash buffer Y2 (4,8 ml), elution buffer EB (12 ml), RNase A (280 μl), column S (50 ea), collection tube (50 ea), elution tube (50 ea).